missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediamine, TRC

Product Code. 30424926
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424926 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424926 Supplier Toronto Research Chemicals Supplier No. TRCP319840500MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Phenylenediamine
CAS 95-54-5
Purity Grade Notes HPLC
Molecular Formula C6 H8 N2
Quantity 500 mg
Synonym o-Phenylenediamine (8CI), 1,2-Diaminobenzene, 1,2-Phenylenediamine, 2-Aminoaniline, C.I. 76010, C.I. Oxidation Base 16, IK 3, IK 3 (amine), NSC 5354, OPDA, Orthamine, o-Aminoaniline, o-Aminophenylamine, o-Benzenediamine, o-Diaminobenzene
InChI Formula InChI=1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2
SMILES Nc1ccccc1N
IUPAC Name benzene-1,2-diamine
Molecular Weight (g/mol) 108.14
Formula Weight 108.0687
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.