missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediacetic acid, 98+%

Product Code. 11471508
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471508 5 g 5g
11481508 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471508 Supplier Thermo Scientific Alfa Aesar Supplier No. A13851.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7500-53-0 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7500-53-0
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004329
InChI Key MMEDJBFVJUFIDD-UHFFFAOYSA-N
Synonym 1,2-phenylenediacetic acid, 2,2'-1,2-phenylene diacetic acid, o-phenylenediacetic acid, o-benzenediacetic acid, 1,2-benzenediacetic acid, 2-2-carboxymethyl phenyl acetic acid, phenylenediacetic acid, acmc-209owx, 1,2-benzenediaceticacid, o-phenylene-diacetic acid
PubChem CID 24123
IUPAC Name 2-[2-(carboxymethyl)phenyl]acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)CC(=O)O

Specifications

Melting Point 150°C to 152°C
Odor Odorless
Quantity 5 g
Beilstein 2616747
Formula Weight 194.19
Percent Purity ≥98%
Chemical Name or Material 1,2-Phenylenediacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.