missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Phenyl-1,3-thiazol-4-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10502473
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10502473

Brand: Thermo Scientific Maybridge SPB03431EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10045-52-0
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.259
MDL Number MFCD00068112
InChI Key ZOOGZFPRAKXWKI-UHFFFAOYSA-N
Synonym 1-2-phenylthiazol-4-yl ethanone, 1-2-phenyl-1,3-thiazol-4-yl ethan-1-one, 1-2-phenyl-1,3-thiazol-4-yl ethanone, 4-acetyl-2-phenylthiazole, 1-2-phenyl-1,3-thiazol-4-yl-1-ethanone, ethanone, 1-2-phenyl-4-thiazolyl, 4-acetyl-2-phenyl-1,3-thiazole, maybridge1_008631, buttpark 15357-18, 2-phenyl-4-acetylthiazole
PubChem CID 604819
IUPAC Name 1-(2-phenyl-1,3-thiazol-4-yl)ethanone
SMILES CC(=O)C1=CSC(=N1)C2=CC=CC=C2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 203.26
Percent Purity 95%
Chemical Name or Material 1-(2-Phenyl-1, 3-thiazol-4-yl)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.