missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Phenoxyphenyl)methanamine hydrochloride, ≥97%, Thermo Scientific™

Product Code. 10071934
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 31963-35-6
Molecular Formula C13H14ClNO
Molecular Weight (g/mol) 235.711
MDL Number MFCD08271962
InChI Key USRYZTSPSJXQFU-UHFFFAOYSA-N
Synonym 1-2-phenoxyphenyl methanamine hydrochloride, 2-phenoxybenzylamine hydrochloride, 2-phenoxyphenyl methanamine hydrochloride, 2-phenoxybenzylamine hcl, 2-phenoxy-benzylamine hydrochloride, 2-phenoxyphenyl methylamine, chloride, 1-2-phenoxyphenyl methanaminehydrochloride, 2-aminomethyl diphenyl ether hydrochloride, 1-2-phenoxyphenyl methanamine-hydrogen chloride 1/1
PubChem CID 17749840
IUPAC Name (2-phenoxyphenyl)methanamine;hydrochloride
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 235.71
Percent Purity 97%
Chemical Name or Material 1-(2-Phenoxyphenyl)methanamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'