missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Phenoxyphenyl)ethanone, 97%, Thermo Scientific™

Product Code. 10006964
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10006964 5 g 5g
10759035 250 mg 250mg
10552154 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10006964 Supplier Thermo Scientific Maybridge Supplier No. CC49928DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 26388-13-6
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD08271961
InChI Key KPBCVVSDGJBODL-UHFFFAOYSA-N
Synonym 1-2-phenoxyphenyl ethanone, 2-acetyldiphenyl ether, 1-acetyl-2-phenoxybenzene, ethanone, 1-2-phenoxyphenyl
PubChem CID 10703750
IUPAC Name 1-(2-phenoxyphenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1OC1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 1-(2-Phenoxyphenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.