missing translation for 'onlineSavingsMsg'
Learn More

1,2-Pentanediol, 96%

Product Code. 15284614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15284614 250 mL Glass bottle 250mL
15264614 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15284614 Supplier Thermo Scientific Acros Supplier No. 123292500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5343-92-0 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5343-92-0
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00010736
InChI Key WCVRQHFDJLLWFE-UHFFFAOYNA-N
Synonym 1,2-pentanediol, 1,2-dihydroxypentane, 1,2-pentanediol, 2r, acmc-20mbh5, acmc-1axdb, 3-01-00-02191 beilstein handbook reference, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa
PubChem CID 93000
IUPAC Name pentane-1,2-diol
SMILES CCCC(O)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9710g/mL
Boiling Point 206.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)2CH(OH)CH2OH
Refractive Index 1.4387 to 1.4407
Quantity 250 mL
Beilstein 01,II,548
Specific Gravity 0.971
Solubility Information Solubility in water: miscible.
Formula Weight 104.15
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1, 2-Pentanediol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Pentanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Pentane-1,2-diol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.