missing translation for 'onlineSavingsMsg'
Learn More

1,2-O-Isopropylidene-alpha-D-glucofuranose

Product Code. 11426770
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426770 25 g 25g
11416770 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426770 Supplier Thermo Scientific Alfa Aesar Supplier No. L10688.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18549-40-1 | C9H16O6 | 220.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18549-40-1
Molecular Formula C9H16O6
Molecular Weight (g/mol) 220.221
MDL Number MFCD00063244
InChI Key BGGCXQKYCBBHAH-OZRXBMAMSA-N
Synonym 1,2-o-isopropylidene-alpha-d-glucofuranose, monoacetone glucose, 1,2-o-isopropylidene-d-glucofuranose, monoacetone-d-glucose, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, 1r-1-3ar,5r,6s,6ar-6-hydroxy-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, glucofuranose, 1,2-o-isopropylidene-, alpha-d, pubchem23862, micro yen+/-ui feminineaeiinic, 1,2-o-isopropylidene--d-glucofuranose
PubChem CID 87704
IUPAC Name (1R)-1-[(3aR,5R,6S,6aR)-6-hydroxy-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
SMILES CC1(OC2C(C(OC2O1)C(CO)O)O)C

Specifications

Melting Point 159°C to 160°C
Quantity 25 g
Beilstein 82670
Formula Weight 220.22
Chemical Name or Material 1,2-O-Isopropylidene-alpha-D-glucofuranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.