missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine, 97%, Thermo Scientific™

Product Code. 11390517
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390517 25 g 25g
10202822 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390517 Supplier Thermo Scientific Maybridge Supplier No. BTB03244EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 58315-38-1
Molecular Formula C11H12F3N3O2
Molecular Weight (g/mol) 275.231
MDL Number MFCD00052613
InChI Key YOBUPGXTLFRIJD-UHFFFAOYSA-N
Synonym 1-2-nitro-4-trifluoromethyl phenyl piperazine, 1-2-nitro-4-trifluoromethylphenyl-piperazine, n-2-nitro-4-trifluoromethyl phenyl piperazine, piperazine, 1-2-nitro-4-trifluoromethyl phenyl, 2-nitro-4-trifluoromethyl phenyl piperazine, 1-2-nitro-4-trifluoromethylphenyl piperazine, 1-2-nitro-4-trifluoromethyl-phenyl-piperazine, piperazine,1-2-nitro-4-trifluoromethyl phenyl
PubChem CID 2771407
IUPAC Name 1-[2-nitro-4-(trifluoromethyl)phenyl]piperazine
SMILES C1CN(CCN1)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 275.23
Percent Purity 97%
Chemical Name or Material 1-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.