missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthoquinone, 95%, Tech.

Product Code. 10229323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10229323 5 g Glass bottle 5g
10138003 1 g Glass bottle 1g
10553731 10 g Glass bottle 10g
10553921 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10229323 Supplier Thermo Scientific Acros Supplier No. 171230050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 524-42-5 | C10H6O2 | 158.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-42-5
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.16
MDL Number MFCD00001698
InChI Key KETQAJRQOHHATG-UHFFFAOYSA-N
Synonym 1,2-naphthoquinone, 1,2-naphthalenedione, o-naphthoquinone, beta-naphthoquinone, 1,2-naphthaquinone, 1,2-naftochinon, .beta.-naphthoquinone, 1,2-naftochinon czech, ccris 1558, 1,2-dihydronaphthalene-1,2-dione
PubChem CID 10667
ChEBI CHEBI:34055
IUPAC Name naphthalene-1,2-dione
SMILES O=C1C=CC2=CC=CC=C2C1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 136.0°C to 141.0°C
Color Colorless or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 07, 709
Fieser 01,713
Merck Index 15, 6479
Solubility Information Solubility in water: negligible. Other solubilities: soluble in 5% naoh,5% nahco3 and conc. h2so4,soluble in alcohol,benzene and ether
Formula Weight 158.16
Percent Purity 95%
Grade Technical
Physical Form Crystalline Mass or Waxy Solid
Chemical Name or Material 1, 2-Naphthoquinone, tech., 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Naphthoquinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.