missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%

Product Code. 10075770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10075770 25 g Glass bottle 25g
10578833 10 g Glass bottle 10g
10269410 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10075770 Supplier Thermo Scientific Acros Supplier No. 415400250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 521-24-4 | C10H5NaO5S | 260.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-24-4
Molecular Formula C10H5NaO5S
Molecular Weight (g/mol) 260.19
MDL Number MFCD00001700
InChI Key UBLXEEBHYISRFM-UHFFFAOYSA-M
Synonym 1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245
PubChem CID 516996
IUPAC Name sodium;3,4-dioxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 289.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 25 g
Beilstein 02, 330
Merck Index 15, 8777
Solubility Information Solubility in water: 33.3g/L (20°C)
Formula Weight 260.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2-Naphthoquinone-4-sulfonic acid sodium salt, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Naphthoquinone-4-sulfonic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.