missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%

Product Code. 11478753
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478753 25 g 25g
11488753 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478753 Supplier Thermo Scientific Alfa Aesar Supplier No. A10507.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 521-24-4 | C10H5NaO5S | 260.195 g/mol

Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.

Solubility
Soluble in water and acetone. Slightly soluble in alcohol and ethanol. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-24-4
Molecular Formula C10H5NaO5S
Molecular Weight (g/mol) 260.195
MDL Number MFCD00001700
InChI Key UBLXEEBHYISRFM-UHFFFAOYSA-M
Synonym 1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245
PubChem CID 516996
IUPAC Name sodium;3,4-dioxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼289°C (decomposition)
Quantity 25 g
Beilstein 4169788
Merck Index 14,8646
Solubility Information Soluble in water and acetone; Slightly soluble in alcohol and ethanol; Insoluble in chloroform.
Formula Weight 260.2
Percent Purity 97%
Chemical Name or Material 1,2-Naphthoquinone-4-sulfonic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.