Learn More
Ratiolab™ 1.2 mL Micro-Tube Refill
Click to view available options
Holds:
Microtubes (96 x 1.2 mL)
Unit Size:
Pack of 4800
Description
- 96-well refill
- Coded micro-tubes
- Convenient handling and reduced waste
- Pressure-tested tightness
- Standard 8 x 12 SBS microplate format
Specifications
Specifications
| Array | 8 x 12 |
| Holds | Microtubes (96 x 1.2 mL) |
| For Use With (Application) | Fast and convenient rack loading |
| For Use With (Equipment) | Micro-Rack systems |
| Quantity | 4800/pk |