missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methylphenyl)piperazine, 98%

Product Code. p-3211195 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10290353 1 g Glass bottle 1g
10015752 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10290353 Supplier Thermo Scientific Acros Supplier No. 382130010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39512-51-1 | C11H16N2 | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39512-51-1
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00040729
InChI Key WICKLEOONJPMEQ-UHFFFAOYSA-N
Synonym 1-2-methylphenyl piperazine, 1-o-tolyl piperazine, 4-2-methylphenyl piperazine, 1-o-tolylpiperazine, piperazine, 1-2-methylphenyl, 1-o-tolyl-piperazine, 1-2-methylphenyl-piperazine, o-tolylpiperazine, 2-methylphenylpiperazine, chembl9746
PubChem CID 91965
IUPAC Name 1-(2-methylphenyl)piperazine
SMILES CC1=CC=CC=C1N2CCNCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 176.26
Percent Purity 98%
Physical Form Low Melting Mass, Crystals or Crystalline Powder
Chemical Name or Material 1-(2-Methylphenyl)piperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(2-Methylphenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.