missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyphenyl)piperazine, 98+%, Thermo Scientific™

Product Code. 10215370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215370 25 g Glass bottle 25g
10202172 5 g Glass bottle 5g
13418338 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215370 Supplier Thermo Scientific Acros Supplier No. 126040250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 35386-24-4
Molecular Formula C11H18Cl2N2O
Molecular Weight (g/mol) 265.18
MDL Number MFCD00005958
InChI Key ZGWQDMTYAQEMHA-UHFFFAOYSA-N
Synonym 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl
PubChem CID 1346
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 35.0°C to 38.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 130.0°C to 133.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. (o-anisidine) (GC)
Packaging Glass bottle
Refractive Index 1.5745 to 1.5765
Quantity 25 g
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
IUPAC Name dihydrogen 1-(2-methoxyphenyl)piperazine dichloride
Formula Weight 192.26
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(2-Methoxyphenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.