missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methoxyphenyl)piperazine, 98+%, Thermo Scientific™
£29.09 - £274.01
Chemical Identifiers
CAS | 35386-24-4 |
---|---|
Molecular Formula | C11H18Cl2N2O |
Molecular Weight (g/mol) | 265.18 |
MDL Number | MFCD00005958 |
InChI Key | ZGWQDMTYAQEMHA-UHFFFAOYSA-N |
Synonym | 1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl |
PubChem CID | 1346 |
SMILES | [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10202172
|
Thermo Scientific Acros
126040050 |
5 g | Glass bottle | ||||||
10215370
|
Thermo Scientific Acros
126040250 |
25 g | Glass bottle | ||||||
13418338
|
Thermo Scientific Acros
126041000 |
100 g | Glass bottle | ||||||
Description
Chemical Identifiers
35386-24-4 | |
265.18 | |
ZGWQDMTYAQEMHA-UHFFFAOYSA-N | |
1346 |
C11H18Cl2N2O | |
MFCD00005958 | |
1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl | |
[H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1 |
Specifications
35386-24-4 | |
100.0 | |
Colorless to Yellow | |
130.0°C to 133.0°C (0.1 mmHg) | |
Authentic | |
C11H18Cl2N2O | |
MFCD00005958 | |
1-2-methoxyphenyl piperazine, 1-2-methoxy-phenyl-piperazine, 1-o-methoxyphenyl piperazine, 2-mpp, 1-2-methoxyphenyl-piperazine, n-2-methoxyphenyl piperazine, unii-81njo1330a, 1-2-methoxy phenyl piperazine, 2-methoxyphenylpiperazine, piperazine, 1-2-methoxyphenyl | |
ZGWQDMTYAQEMHA-UHFFFAOYSA-N | |
1-(2-methoxyphenyl)piperazine | |
1346 | |
98+% | |
1-(2-Methoxyphenyl)piperazine, 98+% |
98.0 | |
35.0°C to 38.0°C | |
1.0900g/mL | |
>110°C | |
1% max. (o-anisidine) (GC) | |
1.5745 to 1.5765 | |
1.09 | |
Solubility in water: insoluble | |
[H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1N1CCNCC1 | |
265.18 | |
192.26 | |
Liquid After Melting |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 252-537-7
missing translation for 'tsca' : TSCA
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-(2-Methoxyphenyl)piperazine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only