missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyethyl)piperazine, 98%

Product Code. 10754943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10754943 5 g Glass bottle 5g
10252762 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10754943 Supplier Thermo Scientific Acros Supplier No. 367110050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13484-40-7 | C7H16N2O | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13484-40-7
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.21
MDL Number MFCD00191214
InChI Key BMEMBBFDTYHTLH-UHFFFAOYSA-N
Synonym 1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-2-methoxy-ethyl-piperazine, piperazine, 1-2-methoxyethyl, 1-methoxy-2-piperazinylethane, pubchem14209, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, 2-methoxyethyl piperazine, n-methoxyethylylpiperazine
PubChem CID 2734638
IUPAC Name 1-(2-methoxyethyl)piperazine
SMILES COCCN1CCNCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 193°C to 194°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.475
Quantity 5 g
Specific Gravity 0.97
Formula Weight 144.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(2-Methoxyethyl)piperazine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2-Methoxyethyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.