missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine Dihydrochloride, TRC

Product Code. 30390446
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390446 50 mg -
30398912 500 mg -
30394040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390446 Supplier Toronto Research Chemicals Supplier No. TRCI87444850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185167-55-8
Molecular Formula C8 H14 N2 S . 2 Cl H
Molecular Weight (g/mol) 243.2
Synonym 2-Isopropyl-4-(methylaminomethyl)thiazole dihydrochloride, 4-Thiazolemethanamine, N-methyl-2-(1-methylethyl)-, hydrochloride (1:2), 1-[2-(1-Methylethyl)thiazol-4-yl]-N-methylmethanamine dihydrochloride
IUPAC Name N-methyl-1-(2-propan-2-yl-1,3-thiazol-4-yl)methanamine;dihydrochloride
SMILES Cl.Cl.CNCc1csc(n1)C(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H14N2S.2ClH/c1-6(2)8-10-7(4-9-3)5-11-8;;/h5-6,9H,4H2,1-3H3;2*1H
Formula Weight 242.0411
Chemical Name or Material 1-(2-Isopropylthiazol-4-yl)-N-methylmethanamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.