missing translation for 'onlineSavingsMsg'
Learn More

1,2-Indanedione, 95%

Product Code. 15698695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15698695 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15698695 Supplier Thermo Scientific Acros Supplier No. 460920010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16214-27-0 | C9H6O2 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16214-27-0
Melting Point 117.0°C to 124.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C9H6O2
Quantity 1 g
Synonym 1h-indene-1,2 3h-dione, 1,2-indanedione, indan-1,2-dione, 1,2-indandione, indandione, 2,3-dihydro-1h-indene-1,2-dione, indane dione, acmc-20a7vb, d01xgp
InChI Key WFFZGYRTVIPBFN-UHFFFAOYSA-N
SMILES C1C2=CC=CC=C2C(=O)C1=O
IUPAC Name 3H-indene-1,2-dione
Molecular Weight (g/mol) 146.15
PubChem CID 123358
Formula Weight 146.15
Percent Purity 95%
Chemical Name or Material 1,2-Indanedione
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Indanedione, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Indanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.