missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyphenyl)ethanone (2-Hydroxyacetophenone), Mikromol™

Product Code. 30050094
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050094 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050094 Supplier Mikromol Supplier No. MM0042.13
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(2-Hydroxyphenyl)ethanone (2-Hydroxyacetophenone)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-Hydroxyphenyl)ethanone (2-Hydroxyacetophenone)
CAS 118-93-4
Analyte or Component Names 1-(2-Hydroxyphenyl)ethanone (2-Hydroxyacetophenone)
Shipping Condition Room Temperature
Molecular Formula C8 H8 O2
Certifications/Compliance ISO 17025
Synonym 1-(2-Hydroxyphenyl)ethanone, Paracetamol Imp. I (EP), 2-Hydroxyacetophenone, Paracetamol Impurity I
InChI Formula InChI=1S/C8H8O2/c1-6(9)7-4-2-3-5-8(7)10/h2-5,10H,1H3
SMILES CC(=O)c1ccccc1O
IUPAC Name 1-(2-hydroxyphenyl)ethanone
Molecular Weight (g/mol) 136.15
Quantity 100 mg
Formula Weight 136.0524 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.