missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)lepidinium Bromide, TRC

Product Code. 30389611
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389611 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389611 Supplier Toronto Research Chemicals Supplier No. TRCH8252155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-Hydroxyethyl)lepidinium Bromide
CAS 26468-13-3
Molecular Formula C12H14BrNO
Quantity 5 mg
Synonym N-(2-Hydroxyethyl)-4-methylquinolinium Bromide, 1-(2-Hydroxyethyl)lepidinium Bromide
InChI Formula InChI=1S/C12H14NO.BrH/c1-10-6-7-13(8-9-14)12-5-3-2-4-11(10)12;/h2-7,14H,8-9H2,1H3;1H/q+1;/p-1
SMILES CC1=CC=[N+](C2=CC=CC=C12)CCO.[Br-]
IUPAC Name 2-(4-methylquinolin-1-ium-1-yl)ethanol;bromide
Molecular Weight (g/mol) 268.15
Formula Weight 267.0259
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.