missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl)-4-methylpiperazine, 98%

Product Code. 10065163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10065163 1 g Glass bottle 1g
10539803 250 mg Glass bottle 250mg
10227233 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10065163 Supplier Thermo Scientific Acros Supplier No. 384830010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5464-12-0 | C7H16N2O | 144.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5464-12-0
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD03840691
InChI Key QHTUMQYGZQYEOZ-UHFFFAOYSA-N
Synonym 1-2-hydroxyethyl-4-methylpiperazine, 2-4-methylpiperazin-1-yl ethanol, 1-piperazineethanol, 4-methyl, 2-4-methylpiperazin-1-yl ethan-1-ol, 1-n-hydroxyethyl-4-methyl piperazine, 2-4-methyl-piperazin-1-yl-ethanol, 1-2-hydroxyethyl-4-methyl-piperazine, 4-methylpiperazine-1-ethanol, 2-4-methylpiperazinyl ethan-1-ol, 2-4-methyl-1-piperazinyl ethanol
PubChem CID 231184
IUPAC Name 2-(4-methylpiperazin-1-yl)ethanol
SMILES CN1CCN(CC1)CCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 54°C to 55°C (0.5 mbar)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.487
Quantity 1 g
Formula Weight 144.22
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 1-(2-Hydroxyethyl)-4-methylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.