missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Hydroxy-4,6-dimethoxy-3-(1-oxopropoxy)phenyl]-1-propanone, TRC

Product Code. 30391377
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391377 100 mg 100mg
30404614 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391377 Supplier Toronto Research Chemicals Supplier No. TRCH941455100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94190-88-2
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.29
IUPAC Name 2-hydroxy-4,6-dimethoxy-3-propionylphenyl propionate
SMILES CCC(C1=C(OC)C=C(OC)C(OC(CC)=O)=C1O)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H18O6/c1-5-8(15)12-9(18-3)7-10(19-4)14(13(12)17)20-11(16)6-2/h7,17H,5-6H2,1-4H3
Formula Weight 282.11
Chemical Name or Material 1-[2-Hydroxy-4,6-dimethoxy-3-(1-oxopropoxy)phenyl]-1-propanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.