missing translation for 'onlineSavingsMsg'
Learn More

1,2-Hexanediol, TRC

Product Code. 30405453
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405453 100 g
30405452 10 g
30391414 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405453 Supplier Toronto Research Chemicals Supplier No. TRCH294245100G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6920-22-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.17
Synonym (±)-Hexane-1,2-diol, 1,2-Dihydroxyhexane, 1,2-Hexyleneglycol, 5,6-Dihydroxyhexane, DL-1,2-Hexanediol, KMO 6
IUPAC Name hexane-1,2-diol
SMILES CCCCC(O)CO

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H14O2/c1-2-3-4-6(8)5-7/h6-8H,2-5H2,1H3
Formula Weight 118.0994
Chemical Name or Material 1,2-Hexanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.