missing translation for 'onlineSavingsMsg'
Learn More

[1-(2-Furylmethyl)piperid-4-yl]methanol, 90%, Thermo Scientific™

Product Code. 10255464
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255464 1 g Amber Glass Bottle 1g
10749215 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255464 Supplier Thermo Scientific Maybridge Supplier No. CC68509DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930111-13-0
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.262
MDL Number MFCD09879982
InChI Key YBANPRCPQURNFY-UHFFFAOYSA-N
Synonym 1-2-furylmethyl piperid-4-yl methanol, 1-furan-2-ylmethyl piperidin-4-yl methanol, 1-furan-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol,1-2-furanylmethyl, 1-2-furylmethyl-4-piperidyl methan-1-ol, 1-2-furylmethyl piperidin-4-yl methanol hydrochloride, 4-piperidinemethanol, 1-2-furanylmethyl-, hydrochloride
PubChem CID 24229765
IUPAC Name [1-(furan-2-ylmethyl)piperidin-4-yl]methanol
SMILES C1CN(CCC1CO)CC2=CC=CO2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 195.26
Percent Purity 90%
Chemical Name or Material [1-(2-Furylmethyl)piperid-4-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.