missing translation for 'onlineSavingsMsg'
Learn More

[1-(2-Furylmethyl)piperid-4-yl]methanol, 90%, Thermo Scientific™

Product Code. p-3586549
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10749215

Brand: Thermo Scientific Maybridge CC68509CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930111-13-0
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.262
MDL Number MFCD09879982
InChI Key YBANPRCPQURNFY-UHFFFAOYSA-N
Synonym 1-2-furylmethyl piperid-4-yl methanol, 1-furan-2-ylmethyl piperidin-4-yl methanol, 1-furan-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol,1-2-furanylmethyl, 1-2-furylmethyl-4-piperidyl methan-1-ol, 1-2-furylmethyl piperidin-4-yl methanol hydrochloride, 4-piperidinemethanol, 1-2-furanylmethyl-, hydrochloride
PubChem CID 24229765
IUPAC Name [1-(furan-2-ylmethyl)piperidin-4-yl]methanol
SMILES C1CN(CCC1CO)CC2=CC=CO2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 195.26
Percent Purity 90%
Chemical Name or Material [1-(2-Furylmethyl)piperid-4-yl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.