missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Furylmethyl)-1H-pyrrole-2-carbaldehyde, TRC

Product Code. 30391553
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391553 500 mg
30398895 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391553 Supplier Toronto Research Chemicals Supplier No. TRCF701585500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13788-32-4
Molecular Formula C15 H13 N O4
Molecular Weight (g/mol) 271.268
Synonym 1-Furfuryl-pyrrole-2-carboxaldehyde, 1-Furfuryl-2-formylpyrrole, 1-Furfurylpyrrole-2-carboxaldehyde, 2-Formyl-N-furfurylpyrrole
IUPAC Name 5-[(E)-2-nitroethenyl]-2-phenylmethoxyphenol
SMILES Oc1cc(\C=C\[N+](=O)[O-])ccc1OCc2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H13NO4/c17-14-10-12(8-9-16(18)19)6-7-15(14)20-11-13-4-2-1-3-5-13/h1-10,17H,11H2/b9-8+
Formula Weight 271.084
Chemical Name or Material 1-(2-Furylmethyl)-1H-pyrrole-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.