missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Furyl)ethanol, TRC

Product Code. 30390641
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390641 5 g
30390498 1 g
30394487 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390641 Supplier Toronto Research Chemicals Supplier No. TRCF8652605G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4208-64-4
Molecular Formula C6 H8 O2
Molecular Weight (g/mol) 112.126
Synonym (±)-1-(2-Furyl)ethanol, 1-(2-Furanyl)ethanol, 1-(2-Furyl)-1-ethanol, 1-(2-Furyl)ethanol, 1-(2'-Furyl)ethanol, 2-(1-Hydroxyethyl)furan, alpha-Methyl-2-furanmethanol, alpha-Methylfurfuryl Alcohol, alpha-Methyl-2-furanmethanol
IUPAC Name 1-(furan-2-yl)ethanol
SMILES CC(O)c1occc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H8O2/c1-5(7)6-3-2-4-8-6/h2-5,7H,1H3
Formula Weight 112.052
Chemical Name or Material 1-(2-Furyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.