missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Furoyl)piperazine, 98%, Thermo Scientific™

Product Code. 11422798
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422798 500 g 500g
11402798 25 g 25g
11412798 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422798 Supplier Thermo Scientific Alfa Aesar Supplier No. A16773.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40172-95-0
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00038831
InChI Key SADPINFEWFPMEA-UHFFFAOYSA-N
Synonym 1-2-furoyl piperazine, 1-furan-2-carbonyl piperazine, furan-2-yl piperazin-1-yl methanone, n-2-furoyl piperazine, unii-8aux72ll1p, 1-2-furoyl-piperazine, furan-2-yl-piperazin-1-yl-methanone, piperazine, 1-2-furanylcarbonyl, 8aux72ll1p
PubChem CID 550206
IUPAC Name furan-2-yl(piperazin-1-yl)methanone
SMILES C1CN(CCN1)C(=O)C2=CC=CO2

Specifications

Melting Point 66°C to 70°C
Boiling Point 137°C to 140°C (0.1 mmHg)
Quantity 500 g
Beilstein 880196
Sensitivity Air Sensitive
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material 1-(2-Furoyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.