missing translation for 'onlineSavingsMsg'
Learn More

1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™

Product Code. 10180842
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10180842 1 g 1g
10671883 10 g 10g
10470944 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10180842 missing translation for 'mfr' Thermo Scientific Maybridge missing translation for 'supplierNo' SB01820DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 37989-92-7
Molecular Formula C15H13FO
Molecular Weight (g/mol) 228.266
MDL Number MFCD00236612
InChI Key ZXIQZWXVSBJPTC-UHFFFAOYSA-N
Synonym 1-2'-fluoro-1,1'-biphenyl-4-yl propan-1-one, 1-2'-fluoro 1,1'-biphenyl-4-yl propan-1-one, 1-4-2-fluorophenyl phenyl propan-1-one, 4'-2-fluorophenyl propiophenone, 4-2-fluorophenyl propiophenone, 1-propanone,1-2'-fluoro 1,1'-biphenyl-4-yl, maybridge3_006207, 2-fluoro-4'-propanoylbiphenyl
PubChem CID 2737490
IUPAC Name 1-[4-(2-fluorophenyl)phenyl]propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula FC6H4C6H4C(O)CH2CH3
Quantity 1 g
Formula Weight 228.26
Percent Purity 97%
Chemical Name or Material 1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.