missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanediyl Bismethanethiosulfonate, TRC

Product Code. 30371288
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371288 10 mg
30377826 25 mg
30380842 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371288 Supplier Toronto Research Chemicals Supplier No. TRCE89035010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-95-8
Molecular Formula C4 H10 O4 S4
Molecular Weight (g/mol) 250.38
IUPAC Name 1,2-bis(methylsulfonylsulfanyl)ethane
SMILES CS(=O)(=O)SCCSS(=O)(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H10O4S4/c1-11(5,6)9-3-4-10-12(2,7)8/h3-4H2,1-2H3
Formula Weight 249.9462
Percent Purity >95%
Chemical Name or Material 1,2-Ethanediyl Bismethanethiosulfonate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.