missing translation for 'onlineSavingsMsg'
Learn More

1,2-Ethanedisulfonic Acid Diethyl Ester, TRC

Product Code. 30388740
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388740 5 g 5g
30376770 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388740 Supplier Toronto Research Chemicals Supplier No. TRCE6765155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500692-41-1
Molecular Formula C6H14O6S2
Molecular Weight (g/mol) 246.3
Synonym NSC 70888, 1,2-Ethanedisulfonic Acid 1,2-Diethyl Ester
IUPAC Name diethyl ethane-1,2-disulfonate
SMILES CCOS(=O)(=O)CCS(=O)(=O)OCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H14O6S2/c1-3-11-13(7,8)5-6-14(9,10)12-4-2/h3-6H2,1-2H3
Formula Weight 246.0232
Chemical Name or Material 1,2-Ethanedisulfonic Acid Diethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.