missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxybutane, TRC

Product Code. 30384127
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384127 50 g -
30376892 1 g -
30377338 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384127 Supplier Toronto Research Chemicals Supplier No. TRCE58910550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-88-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
Synonym 2-Ethyl-oxirane, (±)-2-Ethyloxirane, (±)-Ethyloxirane, 1,2-Butene Oxide, 1,2-Butylene Epoxide, 1,2-Butylene Oxide, 1,2-Epoxybutane, 1-Butene Oxide, 1-Butylene Oxide, 2-Ethyloxirane, Butene 1,2-epoxide, Butylene Oxide, DL-1,2-Epoxybutane, Epoxybutane, Ethylethylene Oxide, Ethyloxirane, NSC 24240, alpha-Butylene Oxide
IUPAC Name 2-ethyloxirane
SMILES CCC1CO1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H8O/c1-2-4-3-5-4/h4H,2-3H2,1H3
Formula Weight 72.0575
Chemical Name or Material 1,2-Epoxybutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.