missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-5-hexene, 98%

Product Code. 10221373 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10221373

Brand: Thermo Scientific Acros 372580250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10353-53-4 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10353-53-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00010051
InChI Key MUUOUUYKIVSIAR-UHFFFAOYSA-N
Synonym 1,2-epoxy-5-hexene, diallyl monoxide, 2-3-butenyl oxirane, diallyl monooxide, oxirane, 3-butenyl, 3-butenyloxirane, 1-hexene, 5,6-epoxy, 2-but-3-en-1-yl oxirane, 5,6-epoxyhex-1-ene, ccris 3748
PubChem CID 66314
IUPAC Name 2-but-3-enyloxirane
SMILES C=CCCC1CO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 118°C to 121°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.426
Quantity 25 g
Specific Gravity 0.87
Solubility Information Solubility in water: .. Other solubilities: soluble in aliphatic solvents,halogenated,solvents and alcohols
Formula Weight 98.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Epoxy-5-hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Epoxy-5-hexene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.