missing translation for 'onlineSavingsMsg'
Learn More

1,2-Distearoyl-sn-glycerol, TRC

Product Code. 30369977
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369977 250 mg
30370081 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369977 Supplier Toronto Research Chemicals Supplier No. TRCD493515250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10567-21-2
Molecular Formula C39H76O5
Molecular Weight (g/mol) 625.02
Synonym Octadecanoic Acid 1,1'-[(1S)-1-(Hydroxymethyl)-1,2-ethanediyl] Ester, Octadecanoic Acid (1S)-1-(Hydroxymethyl)-1,2-ethanediyl Ester, (S)-Octadecanoic Acid 1-(Hydroxymethyl)-1,2-ethanediyl Ester, L-1,2-Di-tearin, 1,2-Di-O-stearoyl-sn-glycerol, 1,2-Distearoyl-sn-glycerol, L-1,2-Distearin, sn-1,2-Distearin
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@H](CO)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3/t37-/m0/s1
Formula Weight 624.5693
Chemical Name or Material 1,2-Distearoyl-sn-glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.