missing translation for 'onlineSavingsMsg'
Learn More

1,2-Distearoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt, TRC

Product Code. 30381909
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Product Code. Quantity
30381909 100 mg
30378764 10 mg
30381699 250 mg
3 options
This item is not returnable. View return policy

Product Code. 30381909

Brand: Toronto Research Chemicals TRCD493585100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200880-42-8
Formula Weight 800.5543
InChI Formula InChI=1S/C42H83O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h39-40,43-44H,3-38H2,1-2H3,(H,47,48);/q;+1/p-1/t39?,40-;/m1./s1
Molecular Formula C42H82NaO10P
Molecular Weight (g/mol) 801.06
Percent Purity >95%
SMILES [Na+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC(O)CO)OC(=O)CCCCCCCCCCCCCCCCC
Synonym Octadecanoic Acid (1R)-1-[[[(2,3-Dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl Ester Monosodium Salt, 1,2-Dioctadecanoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Monosodium Salt
Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt
Grade HPLC

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.