missing translation for 'onlineSavingsMsg'
Learn More

1,2-Distearoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt, TRC

Product Code. 30381699
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381699 250 mg
30381909 100 mg
30378764 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381699 Supplier Toronto Research Chemicals Supplier No. TRCD493585250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200880-42-8
Molecular Formula C42H82NaO10P
Molecular Weight (g/mol) 801.06
Synonym Octadecanoic Acid (1R)-1-[[[(2,3-Dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl Ester Monosodium Salt, 1,2-Dioctadecanoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Monosodium Salt
SMILES [Na+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC(O)CO)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C42H83O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h39-40,43-44H,3-38H2,1-2H3,(H,47,48);/q;+1/p-1/t39?,40-;/m1./s1
Formula Weight 800.5543
Percent Purity >95%
Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.