missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
1,2-Diphenylhydrazine, SPEX CertiPrep™
Shop alle SPEX Certiprep produkter
Klik for at se tilgængelige muligheder
Quantity:
1 mL
Pakningsstørrelse:
1mL
Beskrivelse
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,2-Diphenylhydrazine | 122-66-7 | 1000μg/mL |
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Tekniske data
| Packaging | 2mL amber prescored glass ampule |
| CAS | 122-66-7, 67-56-1 |
| CAS Min % | 0.1, 99.9 |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| CAS Max % | 0.1, 99.9 |
| Molecular Formula | C12H12N2 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI Key | YBQZXXMEJHZYMB-UHFFFAOYSA-N |
| Type | Certified Reference Material |
| SMILES | C1=CC=C(C=C1)NNC2=CC=CC=C2 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- 1,2-Diphenylhydrazine
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 1,2-diphenylhydrazine, methanol
Product intended for laboratory use only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur