missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylethane, 98+%

Product Code. 11457556
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457556 25 g 25g
11467556 100 g 100g
11477556 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457556 Supplier Thermo Scientific Alfa Aesar Supplier No. A14617.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-29-7 | C14H14 | 182.27 g/mol

1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diphenylethane is used as solvent for nitro fibre and in synthesis of other organic chemical products.

Solubility
Soluble in ether, chloroform

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-29-7
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00004796
InChI Key QWUWMCYKGHVNAV-UHFFFAOYSA-N
Synonym 1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
PubChem CID 7647
ChEBI CHEBI:34047
IUPAC Name 2-phenylethylbenzene
SMILES C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 50°C to 53°C
Density 1.013
Boiling Point 284°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 508068
Merck Index 14,1198
Solubility Information Soluble in ether, chloroform
Formula Weight 182.27
Percent Purity ≥98%
Chemical Name or Material 1,2-Diphenylethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.