missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diphenylcyclopropane, cis + trans, 97%

Product Code. 11352736
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352736 2 g 2g
11362736 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352736 Supplier Thermo Scientific Alfa Aesar Supplier No. L08833.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29881-14-9 | C15H14 | 194.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29881-14-9
Molecular Formula C15H14
Molecular Weight (g/mol) 194.277
MDL Number MFCD00040875
InChI Key ZSIYTDQNAOYUNE-UHFFFAOYSA-N
Synonym 1,2-diphenylcyclopropane, 2-phenylcyclopropyl benzene, 1,2-diphenylcyclopropane, cis + trans, benzene, 1,1'-1,2-cyclopropanediyl bis, 1,2-diphenylcyclopropane, trans, acmc-1cckp, trans-1,1'-1,2-cyclopropanediyl bisbenzene, benzene, 1,1'-1,2-cyclopropanediyl bis-, cis, 2-phenylcyclopropyl benzene #, benzene,1,1'-1,2-cyclopropanediyl bis
PubChem CID 70824
IUPAC Name (2-phenylcyclopropyl)benzene
SMILES C1C(C1C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Density 1.02
Boiling Point 175°C to 177°C (14 mmHg)
Refractive Index 1.595
Quantity 2 g
Beilstein 2043299
Formula Weight 194.28
Percent Purity 97%
Chemical Name or Material 1,2-Diphenylcyclopropane, Cis + Trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.