Learn More
1,2-Diphenyl-3,5-pyrazolidinedione, 98%, Thermo Scientific™
Brand: Thermo Scientific Acros 232260010
Code : 29
Additional Details : CAS Number : 2652-77-9 Weight : 0.05100kg
Quantity | 1g |
---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2652-77-9 | |
252.273 | |
XDPKQGKEOCYMQC-UHFFFAOYSA-N | |
17561 | |
C1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3 |
C15H12N2O2 | |
MFCD00179924 | |
1,2-diphenyl-pyrazolidine-3,5-dione, diphenyldiketopyrazolidine, 1,2-diphenyl-3,5-pyrazolidinedione, difenildichetopirazolidina, 1,2-diphenyl-3,5-dioxopyrazolidine, difenildichetopirazolidina italian, 1,2-diphenyl-3,5-dioxopyrazolidin, 3,5-pyrazolidinedione, 1,2-diphenyl, 1,2-diphenyl-3,5-dioxopyrazolidin german, 1,2-diphenyl-1,2-diazolidine-3,5-dione | |
1,2-diphenylpyrazolidine-3,5-dione |
Specifications
1, 2-Diphenyl-3, 5-pyrazolidinedione | |
97.5 | |
Beige-Yellow | |
98% | |
C15H12N2O2 | |
Glass bottle | |
1g | |
C1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3 | |
252.273 | |
252.27 | |
≥97.5% |
2652-77-9 | |
100.0 | |
180°C to 183°C | |
Authentic | |
MFCD00179924 | |
1,2-diphenyl-pyrazolidine-3,5-dione, diphenyldiketopyrazolidine, 1,2-diphenyl-3,5-pyrazolidinedione, difenildichetopirazolidina, 1,2-diphenyl-3,5-dioxopyrazolidine, difenildichetopirazolidina italian, 1,2-diphenyl-3,5-dioxopyrazolidin, 3,5-pyrazolidinedione, 1,2-diphenyl, 1,2-diphenyl-3,5-dioxopyrazolidin german, 1,2-diphenyl-1,2-diazolidine-3,5-dione | |
XDPKQGKEOCYMQC-UHFFFAOYSA-N | |
1,2-diphenylpyrazolidine-3,5-dione | |
17561 | |
Crystalline Powder |