missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dipalmityl-sn-glycero-3-Phosphoethanolamine-N-(cap biotinyl) Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Dipalmityl-sn-glycero-3-Phosphoethanolamine-N-(cap biotinyl) Sodium Salt |
| CAS | 384835-52-3 |
| Molecular Formula | C53H98N4NaO11PS |
| Quantity | 25 mg |
| Synonym | 1,1'-[(1R)-1-[19-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-3-hydroxy-3-oxido-8,15-dioxo-2,4-dioxa-7,14-diaza-3-phosphanonadec-1-yl]-1,2-ethanediyl] ester Hexadecanoic Acid Sodium Salt, Biotinyl Cap PE, C16:0 Biotinyl Cap PE |
| InChI Formula | InChI=1S/C53H99N4O11PS.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-29-37-50(60)65-42-45(68-51(61)38-30-26-24-22-20-18-16-14-12-10-8-6-4-2)43-67-69(63,64)66-41-40-55-49(59)35-28-27-33-39-54-48(58)36-32-31-34-47-52-46(44-70-47)56-53(62)57-52;/h45-47,52H,3-44H2,1-2H3,(H,54,58)(H,55,59)(H,63,64)(H2,56,57,62);/q;+1/p-1/t45-,46+,47+,52+;/m1./s1 |
| SMILES | [Na].CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)OCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)OC(=O)CCCCCCCCCCCCCCC |
| Molecular Weight (g/mol) | 1053.39 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?