missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dipalmitoyl-rac-glycero-3-phosphoethanolamine, TRC

Product Code. 30388210
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388210 500 mg
30388758 50 mg
30376103 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388210 Supplier Toronto Research Chemicals Supplier No. TRCD486820500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5681-36-7
Molecular Formula C37H74NO8P
Molecular Weight (g/mol) 691.96
Synonym 1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl Ester Hexadecanoic Acid, (±)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl Ester Hexadecanoic Acid, DL-1,2-di-Palmitin 2-Aminoethyl Hydrogen Phosphate, dl-1,2-di-Palmitin Phosphate 2-Aminoethyl Ester, 1,2-Dipalmitoyl-3-DL-glycerylphosphorylethanolamine, 1,2-Dipalmitoyl-DL-3-glycerophosphatidylethanolamine, 1,2-Dipalmitoyl-DL-phosphatidylethanolamine, 1,2-Dipalmitoyl-rac-glycerophosphoethanolamine, 1,2-Dipalmitoylphosphatidylethanolamine, DL-Dipalmitoylphosphatidylethanolamine, DL-α-Cephalin dipalmitate, DL-α-Dipalmitoylphosphatidylethanolamine, DPPE, Dipalmitoyl cephalin, Dipalmitoylphosphatidylethanolamine, α-Cephalin, β,γ-dipalmitoyl-, β,γ-Dipalmitoyl-DL-α-cephalin
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)(O)OCCN)OC(=O)CCCCCCCCCCCCCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C37H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34,38H2,1-2H3,(H,41,42)
Chemical Name or Material 1,2-Dipalmitoyl-rac-glycero-3-phosphoethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.