missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dioleoyl-sn-glycero-3-phosphocholine, TRC

Product Code. 30382329
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382329 500 mg
30388490 250 mg
30386389 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382329 Supplier Toronto Research Chemicals Supplier No. TRCD493605500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4235-95-4
Molecular Formula C44H84NO8P
Molecular Weight (g/mol) 786.11
Synonym 3,5,9-Trioxa-4-phosphaheptacos-18-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxo-9-octadecenyl)oxy]-, hydroxide,inner salt, 4-oxide, [R-(Z,Z)]-, 3,5,9-Trioxa-4-phosphaheptacos-18-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-10-oxo-7-[[(9Z)-1-oxo-9-octadecenyl]oxy]-,inner salt, 4-oxide, (7R,18Z)- (9CI), Choline phosphate, 3-ester with L-1,2-diolein (6CI), Choline, hydroxide, dihydrogen phosphate, inner salt, ester withL-1,2-diolein (8CI), Olein, 1,2-di-, L-, dihydrogen phosphate, monoester with choline hydroxide (8CI), 1,2-Dioleoyl-3-sn-phosphatidylcholine, 1,2-Dioleoyl-L-a-lecithin, 1,2-Dioleoyl-sn-glycero-3-phosphocholine, 1,2-Dioleoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioleoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioleoyl-sn-phosphatidylcholine, 3,5,9-Trioxa-4-phosphaheptacos-18-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxo-9-octadecenyl)oxy]-, inner salt, 4-oxide, [R-(Z,Z)]-, DOPC, Dioleoyl L-a-lecithin, Dioleoyl-3-sn-phosphatidylcholine, Dioleoyl-L-a-glycerophosphocholine, Dioleoyl-L-a-glycerophosphorylcholine, Dioleoyl-L-a-phosphatidylcholine, Dioleoylphosphatidylcholine, L-Dioleoyl lecithin, L-a-Di(cis-9-octadecanoyl) lecithin, L-a-Dioleoyl phosphatidylcholine, L-a-Dioleoyllecithin, L-a-Dioleylphosphatidylcholine, PDD 111, sn-3-Dioleoyllecithin
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C44H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,42H,6-19,24-41H2,1-5H3/b22-20-,23-21-/t42-/m1/s1
Formula Weight 785.5935
Percent Purity >95%
Chemical Name or Material 1,2-Dioleoyl-sn-glycero-3-phosphocholine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.