missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dioleoyl-3-palmitoyl-rac-glycerol, TRC

Product Code. 30376897
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30376897

Brand: Toronto Research Chemicals TRCD484200250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dioleoyl-3-palmitoyl-rac-glycerol
CAS 2190-30-9
Purity Grade Notes HPLC
Molecular Formula C55 H102 O6
Quantity 250 mg
Synonym OOP, 9-Octadecenoic acid (9Z)-, 1,1'-[1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl] ester, 9-Octadecenoic acid (9Z)-, 1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl ester (9CI), 9-Octadecenoic acid (Z)-, 1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl ester, Olein, 3-palmito-1,2-di- (7CI,8CI), Palmitin, 2,3-dioleo-1- (8CI), 1,2-Dioleo-3-palmitin, 1,2-Dioleoyl-3-palmitoyl-rac-glycerol, 1,2-Dioleoyl-3-palmitoylglycerol, 1-Palmito-2,3-diolein, 1-Palmito-2-oleo-3-olein, 2-Oleoyl-1-oleoyl-3-palmitin, 3-Palmito-1,2-diolein, Glyceryl 1-palmitate-2,3-dioleate, rac-Glyceryl 1,2-dioleate 3-palmitate
InChI Formula InChI=1S/C55H102O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h25-28,52H,4-24,29-51H2,1-3H3/b27-25-,28-26-
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC Name [3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] (Z)-octadec-9-enoate
Molecular Weight (g/mol) 859.39
Formula Weight 858.7676
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.