missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dioleoyl-3-palmitoyl-rac-glycerol, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 1,2-Dioleoyl-3-palmitoyl-rac-glycerol |
| CAS | 2190-30-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C55 H102 O6 |
| Quantity | 100 mg |
| Synonym | OOP, 9-Octadecenoic acid (9Z)-, 1,1'-[1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl] ester, 9-Octadecenoic acid (9Z)-, 1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl ester (9CI), 9-Octadecenoic acid (Z)-, 1-[[(1-oxohexadecyl)oxy]methyl]-1,2-ethanediyl ester, Olein, 3-palmito-1,2-di- (7CI,8CI), Palmitin, 2,3-dioleo-1- (8CI), 1,2-Dioleo-3-palmitin, 1,2-Dioleoyl-3-palmitoyl-rac-glycerol, 1,2-Dioleoyl-3-palmitoylglycerol, 1-Palmito-2,3-diolein, 1-Palmito-2-oleo-3-olein, 2-Oleoyl-1-oleoyl-3-palmitin, 3-Palmito-1,2-diolein, Glyceryl 1-palmitate-2,3-dioleate, rac-Glyceryl 1,2-dioleate 3-palmitate |
| InChI Formula | InChI=1S/C55H102O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h25-28,52H,4-24,29-51H2,1-3H3/b27-25-,28-26- |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC Name | [3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 859.39 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction