missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimyristoyl-sn-glycero-3-phosphate Sodium Salt, TRC

Product Code. 30384974
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384974 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384974 Supplier Toronto Research Chemicals Supplier No. TRCD47965525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimyristoyl-sn-glycero-3-phosphate Sodium Salt
CAS 80724-31-8
Molecular Formula C31H60NaO8P
Quantity 25 mg
Synonym 1,1'-[(1R)-1-[(Phosphonooxy)methyl]-1,2-ethanediyl] Ester Tetradecanoic Acid Sodium Salt, (1R)-1-[(Phosphonooxy)methyl]-1,2-ethanediyl Ester Tetradecanoic Acid Monosodium Salt, 1,2-Dimyristoyl-sn-glycero-3-phosphate Monosodium Salt, Monosodium 1,2-Dimyristoyl-sn-glycerophosphate
InChI Formula InChI=1S/C31H61O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-30(32)37-27-29(28-38-40(34,35)36)39-31(33)26-24-22-20-18-16-14-12-10-8-6-4-2;/h29H,3-28H2,1-2H3,(H2,34,35,36);/q;+1/p-1/t29-;/m1./s1
SMILES [Na+].CCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)[O-])OC(=O)CCCCCCCCCCCCC
Molecular Weight (g/mol) 614.77
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.