missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-3-hydroxy-4-pyridone, 98+%

Product Code. 10197503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10197503 50 g Glass bottle 50g
10723024 5 g Glass bottle 5g
10702491 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10197503 Supplier Thermo Scientific Acros Supplier No. 278740500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30652-11-0 | C7H9NO2 | 139.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30652-11-0
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.15
MDL Number MFCD00134497
InChI Key TZXKOCQBRNJULO-UHFFFAOYSA-N
Synonym deferiprone, ferriprox, 3-hydroxy-1,2-dimethyl-4 1h-pyridone, 3-hydroxy-1,2-dimethylpyridin-4 1h-one, 1,2-dimethyl-3-hydroxy-4-pyridone, 1,2-dimethyl-3-hydroxypyrid-4-one, 1,2-dimethyl-3-hydroxypyridine-4-one, cp20 chelating agent, unii-2bty8kh53l, ferriprox tn
PubChem CID 2972
ChEBI CHEBI:68554
IUPAC Name 3-hydroxy-1,2-dimethylpyridin-4-one
SMILES CC1=C(C(=O)C=CN1C)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 271°C to 273°C
Color White
pH 6 to 8 (1% soln.)
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 g
Merck Index 15, 2862
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in dmso
Formula Weight 139.15
Percent Purity 99+%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 1, 2-Dimethyl-3-hydroxy-4-pyridone
Show More Show Less
compliance-icons
Product Identifier
  • 1,2-Dimethyl-3-hydroxy-4-pyridone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.