missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine, TRC

Product Code. 30385332
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385332 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385332 Supplier Toronto Research Chemicals Supplier No. TRCD44012025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrantel impurity D
CAS 4271-96-9
Molecular Formula C6 H12 N2
Quantity 25 g
Synonym 1,4,5,6-Tetrahydro-1,2-dimethylpyrimidine, 1,2-Dimethyl-1,4,5,6-tetrahydropyrimidine, 2,3-Dimethyl-3,4,5,6-tetrahydropyrimidine, Pyrantel Embonate Imp. D (EP)
InChI Formula InChI=1S/C6H12N2/c1-6-7-4-3-5-8(6)2/h3-5H2,1-2H3
SMILES CN1CCCN=C1C
IUPAC Name 1,2-dimethyl-5,6-dihydro-4H-pyrimidine
Molecular Weight (g/mol) 112.17
Formula Weight 112.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.