missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Honeywell™

Product Code. 15687840 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Unit Size:
1 L
100mL
2.5 L
20 L
500mL
This item is not returnable. View return policy

Product Code. 15687840

Brand: Honeywell Research Chemicals E27408500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, inhibitor-free,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Melting Point -58°C
Color Colorless
Density 0.867g/mL at 25°C
pH Neutral
Boiling Point 85°C (1013 hPa)
Flash Point −2°C (28.4°F)
Packaging Glass bottle
Linear Formula CH3OCH2CH2OCH3
Quantity 500 mL
UN Number UN2252
Beilstein 1209237
Vapor Density 3.1 (20°C, vs air)
Formula Weight 90.12g/mol
Percent Purity ≥99%
Chemical Name or Material 1,2-Dimethoxyethane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.