missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diiodotetrafluoroethane, 96%, stab. with copper

Product Code. 11434383
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434383 5 g 5g
15427515 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434383 Supplier Thermo Scientific Alfa Aesar Supplier No. A10954.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 354-65-4 | C2F4I2 | 353.825 g/mol

1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodotetrafluoroethane is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-65-4
Molecular Formula C2F4I2
Molecular Weight (g/mol) 353.825
MDL Number MFCD00039401
InChI Key NZXVPCQHQVWOFD-UHFFFAOYSA-N
Synonym 1,2-diiodotetrafluoroethane, 1,2-diiodoperfluoroethane, unii-2z13i2welk, ethane, 1,1,2,2-tetrafluoro-1,2-diiodo, 2z13i2welk, acmc-1cknr
PubChem CID 67720
IUPAC Name 1,1,2,2-tetrafluoro-1,2-diiodoethane
SMILES C(C(F)(F)I)(F)(F)I

Specifications

Melting Point -21°C
Density 2.629
Boiling Point 112°C to 113°C
Flash Point None
Refractive Index 1.495
Quantity 5 g
Beilstein 1740350
Sensitivity Light sensitive
Formula Weight 353.82
Percent Purity 96%
Chemical Name or Material 1,2-Diiodotetrafluoroethane, Stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.